Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTCGTTTTTGTATTTGATTTG[A/G]CAGAGTATGTAGGCTGTGGCTTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606165 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BOLL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BOLL - boule homolog, RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284358.1 | Intron | NP_001271287.1 | ||||
NM_001284361.1 | Intron | NP_001271290.1 | ||||
NM_001284362.1 | Intron | NP_001271291.1 | ||||
NM_033030.5 | Intron | NP_149019.1 | ||||
NM_197970.2 | Intron | NP_932074.1 | ||||
XM_006712715.3 | Intron | XP_006712778.1 | ||||
XM_011511692.2 | Intron | XP_011509994.1 | ||||
XM_011511693.2 | Intron | XP_011509995.1 | ||||
XM_011511694.2 | Intron | XP_011509996.1 | ||||
XM_017004773.1 | Intron | XP_016860262.1 |
Set Membership: |
HapMap |