Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACATTTTGAAATCAGTGTGCCC[A/C]TTAGAACTTTCTTTCCCCTGAAACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603775 MIM: 611351 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNE2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CCNE2 - cyclin E2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_057749.2 | 4467 | Intron | NP_477097.1 | |||
XM_011517366.2 | 4467 | Intron | XP_011515668.1 | |||
XM_017013957.1 | 4467 | Intron | XP_016869446.1 | |||
XM_017013958.1 | 4467 | Intron | XP_016869447.1 | |||
XM_017013959.1 | 4467 | Intron | XP_016869448.1 |
INTS8 - integrator complex subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017864.3 | 4467 | Intron | NP_060334.2 | |||
XM_017013616.1 | 4467 | Intron | XP_016869105.1 | |||
XM_017013617.1 | 4467 | Intron | XP_016869106.1 | |||
XM_017013618.1 | 4467 | UTR 3 | XP_016869107.1 | |||
XM_017013619.1 | 4467 | UTR 3 | XP_016869108.1 |
Set Membership: |
HapMap |