Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCATACCTCTACTGCTCTGTGTC[C/G]CACTACCATGACCAAACCTCCTACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614136 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAPPC8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TRAPPC8 - trafficking protein particle complex 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014939.3 | Intron | NP_055754.2 | ||||
XM_005258233.2 | Intron | XP_005258290.1 | ||||
XM_005258234.2 | Intron | XP_005258291.1 | ||||
XM_005258235.3 | Intron | XP_005258292.1 | ||||
XM_006722420.3 | Intron | XP_006722483.1 | ||||
XM_011525884.2 | Intron | XP_011524186.1 | ||||
XM_017025615.1 | Intron | XP_016881104.1 | ||||
XM_017025616.1 | Intron | XP_016881105.1 | ||||
XM_017025617.1 | Intron | XP_016881106.1 |