Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACGAAGCTCAGGATACCAGACT[A/C]GCGGCGGCCGCCTGGCCCCCGGAAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609353 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC25 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC25 - coiled-coil domain containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304529.1 | 40 | UTR 5 | NP_001291458.1 | |||
NM_001304530.1 | 40 | UTR 5 | NP_001291459.1 | |||
NM_001304532.1 | 40 | UTR 5 | NP_001291461.1 | |||
NM_018246.2 | 40 | UTR 5 | NP_060716.2 | |||
XM_011544570.2 | 40 | Intron | XP_011542872.1 | |||
XM_011544571.2 | 40 | Intron | XP_011542873.1 |
ESCO2 - establishment of sister chromatid cohesion N-acetyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017420.2 | 40 | Intron | NP_001017420.1 | |||
XM_011544421.2 | 40 | Intron | XP_011542723.1 | |||
XM_011544422.2 | 40 | Intron | XP_011542724.1 |
Set Membership: |
HapMap |