Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCATCCTTTATGTAGTTTTTTTT[C/T]AAAGGCAGCTGTTTCTTTAACAACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615288 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DRC1 - dynein regulatory complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145038.4 | Intron | NP_659475.2 | ||||
XM_005264637.3 | Intron | XP_005264694.1 | ||||
XM_005264638.3 | Intron | XP_005264695.1 | ||||
XM_017005271.1 | Intron | XP_016860760.1 |
Set Membership: |
HapMap |