Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTTCCTCCTCCTCCTCATCTTC[A/G]TCTTCTTCTTCTTCAAGCATGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615756 | ||||||||||||||||||||
Literature Links: |
SECISBP2L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SECISBP2L - SECIS binding protein 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193489.1 | 3243 | Silent Mutation | NP_001180418.1 | |||
NM_014701.3 | 3243 | Silent Mutation | NP_055516.2 |