Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCATCCATGCTACGAAAGTCCTC[C/T]ATAGTGGGAACTAGGTCTCCCGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108370 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASNS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASNS - asparagine synthetase (glutamine-hydrolyzing) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178075.1 | Intron | NP_001171546.1 | ||||
NM_001178076.1 | Intron | NP_001171547.1 | ||||
NM_001178077.1 | Intron | NP_001171548.1 | ||||
NM_001673.4 | Intron | NP_001664.3 | ||||
NM_133436.3 | Intron | NP_597680.2 | ||||
NM_183356.3 | Intron | NP_899199.2 |
Set Membership: |
HapMap |