Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAAGCATTCCTTTATTATGTCAA[A/G]CAACCTACCCTTGACCTCTATGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609457 MIM: 151570 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HAL - histidine ammonia-lyase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LTA4H - leukotriene A4 hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000895.2 | Intron | NP_000886.1 | ||||
NM_001256643.1 | Intron | NP_001243572.1 | ||||
NM_001256644.1 | Intron | NP_001243573.1 | ||||
XM_005268871.1 | Intron | XP_005268928.1 | ||||
XM_011538348.1 | Intron | XP_011536650.1 | ||||
XM_011538349.2 | Intron | XP_011536651.1 |