Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAATGCATGCCAGGATAGACAA[C/T]CTACAGTTTCTGCCACCAATAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602841 MIM: 612425 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AOX1 - aldehyde oxidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159.3 | Intron | NP_001150.3 | ||||
XM_011511062.1 | Intron | XP_011509364.1 | ||||
XM_017003946.1 | Intron | XP_016859435.1 | ||||
XM_017003947.1 | Intron | XP_016859436.1 |
SGO2 - shugoshin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |