Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCAGAGACGCCATGTACCGGCTC[A/C]TGTCAGCAGTGACTGCCCGGGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LACTB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LACTB - lactamase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288585.1 | 85 | Missense Mutation | ATG,CTG | M,L 5 | NP_001275514.1 | |
NM_032857.4 | 85 | Missense Mutation | ATG,CTG | M,L 5 | NP_116246.2 | |
NM_171846.3 | 85 | Missense Mutation | ATG,CTG | M,L 5 | NP_741982.1 | |
XM_011521180.1 | 85 | Missense Mutation | ATG,CTG | M,L 5 | XP_011519482.1 |