Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTCTAGACCATTTACCATTAAT[A/G]CAGAGGCTACATTCTTTCAGTCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607534 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
YAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
YAF2 - YY1 associated factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190977.2 | 3777 | UTR 3 | NP_001177906.1 | |||
NM_001190979.2 | 3777 | UTR 3 | NP_001177908.1 | |||
NM_001190980.2 | 3777 | Intron | NP_001177909.1 | |||
NM_001320080.1 | 3777 | UTR 3 | NP_001307009.1 | |||
NM_005748.5 | 3777 | UTR 3 | NP_005739.2 | |||
XM_006719184.3 | 3777 | Intron | XP_006719247.1 | |||
XM_006719185.3 | 3777 | Intron | XP_006719248.1 | |||
XM_011537728.2 | 3777 | Intron | XP_011536030.1 | |||
XM_011537729.2 | 3777 | Intron | XP_011536031.1 | |||
XM_017018669.1 | 3777 | Intron | XP_016874158.1 | |||
XM_017018670.1 | 3777 | Intron | XP_016874159.1 |