Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGAGGATATTAAAGCCTGTTCC[A/C]CTGACTAATTTTCATCAATACCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300550 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHEX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PHEX - phosphate regulating endopeptidase homolog, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000444.5 | Intron | NP_000435.3 | ||||
NM_001282754.1 | Intron | NP_001269683.1 | ||||
XM_011545533.1 | Intron | XP_011543835.1 | ||||
XM_011545536.2 | Intron | XP_011543838.1 | ||||
XM_017029579.1 | Intron | XP_016885068.1 |
PHEX-AS1 - PHEX antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |