Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTCAGTCTAAAGCGAAACATCA[A/G]TAAGTTAATAATAAACCTATCTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615626 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C15orf41 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C15orf41 - chromosome 15 open reading frame 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130010.2 | 2231 | UTR 3 | NP_001123482.1 | |||
NM_001290232.1 | 2231 | Intron | NP_001277161.1 | |||
NM_001290233.1 | 2231 | Intron | NP_001277162.1 | |||
NM_001321756.1 | 2231 | UTR 3 | NP_001308685.1 | |||
NM_001321757.1 | 2231 | UTR 3 | NP_001308686.1 | |||
NM_001321758.1 | 2231 | UTR 3 | NP_001308687.1 | |||
NM_001321759.1 | 2231 | UTR 3 | NP_001308688.1 | |||
NM_001321760.1 | 2231 | UTR 3 | NP_001308689.1 | |||
NM_001321761.1 | 2231 | UTR 3 | NP_001308690.1 | |||
NM_032499.5 | 2231 | UTR 3 | NP_115888.1 | |||
XM_011522111.1 | 2231 | Intron | XP_011520413.1 | |||
XM_017022676.1 | 2231 | UTR 3 | XP_016878165.1 | |||
XM_017022677.1 | 2231 | UTR 3 | XP_016878166.1 |
CSNK1A1P1 - casein kinase 1 alpha 1 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |