Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTCTGGTTAGTAGTTTCCTATC[C/T]CCTTTTGTTACAGGAAAGGAGTCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605656 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXL7 - F-box and leucine rich repeat protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278317.1 | Intron | NP_001265246.1 | ||||
NM_012304.4 | Intron | NP_036436.1 | ||||
XM_011513998.1 | Intron | XP_011512300.1 | ||||
XM_017009262.1 | Intron | XP_016864751.1 |