Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGCACATAACTCAGGGTCTTTAA[C/T]TGCTAACCACCGAAGGTTCAAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162280 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6841 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR6841 - microRNA 6841 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEFL - neurofilament, light polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006158.4 | 2364 | UTR 3 | NP_006149.2 |