Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATCCCTTATTAAAATGCGAATAC[C/T]AAAGCAGGCGAACAGATCATGATAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300520 MIM: 300970 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLDN2 - claudin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MORC4 - MORC family CW-type zinc finger 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085354.2 | 3900 | Intron | NP_001078823.1 | |||
NM_024657.4 | 3900 | Intron | NP_078933.3 | |||
XM_005262190.3 | 3900 | UTR 3 | XP_005262247.1 | |||
XM_006724691.2 | 3900 | UTR 3 | XP_006724754.1 | |||
XM_011531027.2 | 3900 | UTR 3 | XP_011529329.1 | |||
XM_017029844.1 | 3900 | UTR 3 | XP_016885333.1 |