Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCATCTTTTAATAAGTCTTTACC[A/G]TAACTGCCTAGGCCGTTCATCCTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 303630 MIM: 303631 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL4A5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL4A5 - collagen type IV alpha 5 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000495.4 | Intron | NP_000486.1 | ||||
NM_033380.2 | Intron | NP_203699.1 | ||||
XM_011530849.2 | Intron | XP_011529151.2 | ||||
XM_017029259.1 | Intron | XP_016884748.1 | ||||
XM_017029260.1 | Intron | XP_016884749.1 | ||||
XM_017029261.1 | Intron | XP_016884750.1 | ||||
XM_017029262.1 | Intron | XP_016884751.1 | ||||
XM_017029263.1 | Intron | XP_016884752.1 |
COL4A6 - collagen type IV alpha 6 chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |