Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCCAGAATGGAATGAATGGGCT[C/T]TTGGGAAAAGCTGGTCCGACCTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162323 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TACR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TACR1 - tachykinin receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001058.3 | 2078 | UTR 3 | NP_001049.1 | |||
NM_015727.2 | 2078 | Intron | NP_056542.1 |
Set Membership: |
HapMap |