Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGCTGTCTCCTAGGCCTCCCCT[C/T]GCGGGTGCAGAAGATCCGAGCCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611286 | ||||||||||||||||||||
Literature Links: |
RTCA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RTCA - RNA 3'-terminal phosphate cyclase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130841.1 | 238 | Missense Mutation | TCG,TTG | S,L 34 | NP_001124313.1 | |
NM_003729.3 | 238 | Missense Mutation | TCG,TTG | S,L 34 | NP_003720.1 | |
XM_005271298.4 | 238 | UTR 5 | XP_005271355.1 | |||
XM_005271299.4 | 238 | UTR 5 | XP_005271356.1 | |||
XM_017002669.1 | 238 | Intron | XP_016858158.1 |
RTCA-AS1 - RTCA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |