Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTATTTTTCTTCCAAGTTTAGA[C/T]AGCTCTTAAGACAATCCATGGAATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609071 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXW2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXW2 - F-box and WD repeat domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012164.3 | 8184 | UTR 3 | NP_036296.2 | |||
XM_005251910.3 | 8184 | Intron | XP_005251967.1 | |||
XM_005251913.2 | 8184 | Intron | XP_005251970.1 | |||
XM_006717049.3 | 8184 | Intron | XP_006717112.1 | |||
XM_006717051.2 | 8184 | Intron | XP_006717114.1 | |||
XM_011518518.1 | 8184 | Intron | XP_011516820.1 | |||
XM_017014615.1 | 8184 | Intron | XP_016870104.1 | |||
XM_017014616.1 | 8184 | Intron | XP_016870105.1 | |||
XM_017014617.1 | 8184 | Intron | XP_016870106.1 |