Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCCTCCCTCTGACCAAGATCCG[C/G]GGCCTCCTCTGGCCATCTCACCAAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606323 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYFIP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYFIP2 - cytoplasmic FMR1 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037333.2 | 684 | Intron | NP_001032410.1 | |||
NM_001291721.1 | 684 | Intron | NP_001278650.1 | |||
NM_001291722.1 | 684 | Intron | NP_001278651.1 | |||
NM_014376.3 | 684 | Intron | NP_055191.2 | |||
XM_011534516.2 | 684 | Intron | XP_011532818.1 | |||
XM_017009341.1 | 684 | Intron | XP_016864830.1 | |||
XM_017009342.1 | 684 | Intron | XP_016864831.1 |
FNDC9 - fibronectin type III domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001343.3 | 684 | Missense Mutation | CCG,GCG | P,A 166 | NP_001001343.2 |
Set Membership: |
HapMap |