Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAAGTCTTGTCTTTCTAACAAGA[C/T]GAACAGGCAGTGGCTTCTCTTTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLEKHM3 - pleckstrin homology domain containing M3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080475.2 | 7934 | UTR 3 | NP_001073944.1 | |||
XM_005246545.3 | 7934 | UTR 3 | XP_005246602.1 | |||
XM_006712521.3 | 7934 | Intron | XP_006712584.1 | |||
XM_011511160.2 | 7934 | UTR 3 | XP_011509462.1 | |||
XM_011511163.2 | 7934 | Intron | XP_011509465.1 | |||
XM_017004071.1 | 7934 | Intron | XP_016859560.1 | |||
XM_017004072.1 | 7934 | UTR 3 | XP_016859561.1 | |||
XM_017004073.1 | 7934 | UTR 3 | XP_016859562.1 |