Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGTTTTATACAAGCATGCAAAG[C/T]ATATAATAGAATCACATGGAAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137140 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GABRA2 - gamma-aminobutyric acid type A receptor alpha2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005248080.3 | 3241 | UTR 3 | XP_005248137.1 | |||
XM_011513675.1 | 3241 | UTR 3 | XP_011511977.1 | |||
XM_011513676.1 | 3241 | UTR 3 | XP_011511978.1 | |||
XM_017007982.1 | 3241 | UTR 3 | XP_016863471.1 | |||
XM_017007983.1 | 3241 | UTR 3 | XP_016863472.1 |
Set Membership: |
HapMap |