Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATAGGTCCGTAAGCTTCCTCAC[A/G]TCTGAATGCTAATCACTGAATTCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607619 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMD3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FRMD3 - FERM domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244959.1 | Intron | NP_001231888.1 | ||||
NM_001244960.1 | Intron | NP_001231889.1 | ||||
NM_001244961.1 | Intron | NP_001231890.1 | ||||
NM_001244962.1 | Intron | NP_001231891.1 | ||||
NM_174938.5 | Intron | NP_777598.3 | ||||
XM_017014588.1 | Intron | XP_016870077.1 | ||||
XM_017014589.1 | Intron | XP_016870078.1 | ||||
XM_017014590.1 | Intron | XP_016870079.1 | ||||
XM_017014591.1 | Intron | XP_016870080.1 |
LOC102723989 - uncharacterized LOC102723989 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |