Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCTTGTATAAACTTCCACAGGG[C/T]TTTATGTTGATAAACCATCTCTGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMCO5A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMCO5A - transmembrane and coiled-coil domains 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152453.3 | Intron | NP_689666.2 | ||||
XM_005254168.3 | Intron | XP_005254225.1 | ||||
XM_011521262.2 | Intron | XP_011519564.1 | ||||
XM_011521263.1 | Intron | XP_011519565.1 | ||||
XM_011521264.1 | Intron | XP_011519566.1 |