Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGAGTCTCATTGTGACTCTAAC[A/T]CACATAGCCCTTGACTTTCTCTGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611060 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927943 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101927943 - uncharacterized LOC101927943 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETBP1 - SET binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130110.1 | Intron | NP_001123582.1 | ||||
NM_015559.2 | Intron | NP_056374.2 |
Set Membership: |
HapMap |