Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCTCTGTTTTGGTCAAGACTTA[C/T]GTTTAGTCAGCAGCTTAGCAGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610418 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDFY2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDFY2 - WD repeat and FYVE domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052950.3 | 2879 | Intron | NP_443182.1 | |||
XM_011534914.1 | 2879 | Intron | XP_011533216.1 | |||
XM_011534915.2 | 2879 | Intron | XP_011533217.1 | |||
XM_017020386.1 | 2879 | UTR 5 | XP_016875875.1 |