Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCCTGAGGGCAGAGCACAAGCA[A/G]TTGTTAAGAGAGGGGTGAGATTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610418 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDFY2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
WDFY2 - WD repeat and FYVE domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052950.3 | 3673 | Intron | NP_443182.1 | |||
XM_011534914.1 | 3673 | Intron | XP_011533216.1 | |||
XM_011534915.2 | 3673 | Intron | XP_011533217.1 | |||
XM_017020386.1 | 3673 | UTR 5 | XP_016875875.1 |