Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTGGGAATGATTATAACTAACA[C/T]GTAATTTCCCCTACTTTGTGAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP9 - microtubule associated protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039580.1 | 5921 | UTR 3 | NP_001034669.1 | |||
XM_006714306.3 | 5921 | Intron | XP_006714369.1 | |||
XM_011532253.1 | 5921 | UTR 3 | XP_011530555.1 | |||
XM_011532254.2 | 5921 | UTR 3 | XP_011530556.1 | |||
XM_011532255.2 | 5921 | UTR 3 | XP_011530557.1 | |||
XM_017008616.1 | 5921 | Intron | XP_016864105.1 | |||
XM_017008617.1 | 5921 | UTR 3 | XP_016864106.1 | |||
XM_017008618.1 | 5921 | Intron | XP_016864107.1 |