Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGAGTGCTTCTGTAGAAGTGCC[A/C]AGTTCAAACGTTCAGCAACACCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF773 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF773 - zinc finger protein 773 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304334.1 | 548 | Silent Mutation | CCA,CCC | P,P 107 | NP_001291263.1 | |
NM_001304335.1 | 548 | Intron | NP_001291264.1 | |||
NM_001304336.1 | 548 | UTR 3 | NP_001291265.1 | |||
NM_001304337.1 | 548 | Intron | NP_001291266.1 | |||
NM_198542.2 | 548 | Silent Mutation | CCA,CCC | P,P 108 | NP_940944.1 |