Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAAATTGTACTTGGGTGGAGCA[C/T]CAAGAGATTTCTGGATGAGTTCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604076 | ||||||||||||||||||||
Literature Links: |
ZNF134 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF134 - zinc finger protein 134 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF530 - zinc finger protein 530 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321981.1 | 590 | Missense Mutation | CCA,TCA | P,S 91 | NP_001308910.1 | |
NM_020880.4 | 590 | Missense Mutation | CCA,TCA | P,S 124 | NP_065931.3 | |
XM_006723194.1 | 590 | Missense Mutation | CCA,TCA | P,S 124 | XP_006723257.1 | |
XM_011526919.2 | 590 | Missense Mutation | CCA,TCA | P,S 123 | XP_011525221.1 | |
XM_011526921.2 | 590 | Missense Mutation | CCA,TCA | P,S 91 | XP_011525223.1 |