Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAACAACAAGAGAGCTCTTCTGA[C/T]GCTTGTATGGTGGTAAGGAGTTTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602650 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPOP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SPOP - speckle type BTB/POZ protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007226.1 | 2145 | UTR 3 | NP_001007227.1 | |||
NM_001007227.1 | 2145 | UTR 3 | NP_001007228.1 | |||
NM_001007228.1 | 2145 | UTR 3 | NP_001007229.1 | |||
NM_001007229.1 | 2145 | UTR 3 | NP_001007230.1 | |||
NM_001007230.1 | 2145 | UTR 3 | NP_001007231.1 | |||
NM_003563.3 | 2145 | UTR 3 | NP_003554.1 | |||
XM_005257723.4 | 2145 | UTR 3 | XP_005257780.1 | |||
XM_005257724.4 | 2145 | UTR 3 | XP_005257781.1 | |||
XM_017025204.1 | 2145 | UTR 3 | XP_016880693.1 |
Set Membership: |
HapMap |