Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGAATTTACAAGTTGAAAGATGG[A/G]AATATGTTATTCTCGGCAGATGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609880 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KAT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
KAT7 - lysine acetyltransferase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199155.1 | Intron | NP_001186084.1 | ||||
NM_001199156.1 | Intron | NP_001186085.1 | ||||
NM_001199157.1 | Intron | NP_001186086.1 | ||||
NM_001199158.1 | Intron | NP_001186087.1 | ||||
NM_007067.4 | Intron | NP_008998.1 | ||||
XM_011524235.2 | Intron | XP_011522537.1 |
Set Membership: |
HapMap |