Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGGCAAGGCTAGAATATTGAAA[C/T]TATGGCCAACATTGCTTACTTTAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01455 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LINC01455 - long intergenic non-protein coding RNA 1455 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTX3 - metaxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010891.4 | 7587 | UTR 3 | NP_001010891.4 | |||
NM_001167741.1 | 7587 | UTR 3 | NP_001161213.1 | |||
XM_005248495.3 | 7587 | UTR 3 | XP_005248552.1 | |||
XM_017009440.1 | 7587 | UTR 3 | XP_016864929.1 |