Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTACCTGGTGGCTTGATAGGCAG[C/G]ATCCTAAAACAGTACTCAAGGTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611646 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPHKAP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SPHKAP - SPHK1 interactor, AKAP domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142644.1 | Intron | NP_001136116.1 | ||||
NM_030623.3 | Intron | NP_085126.2 | ||||
XM_005246870.4 | Intron | XP_005246927.1 | ||||
XM_006712777.3 | Intron | XP_006712840.1 | ||||
XM_006712778.3 | Intron | XP_006712841.1 | ||||
XM_011511925.2 | Intron | XP_011510227.1 |
Set Membership: |
HapMap |