Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607336 MIM: 600357 | ||||||||||||||||||||
Literature Links: |
BEST4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BEST4 - bestrophin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153274.2 | 1254 | Missense Mutation | CGG,CTG | R,L 402 | NP_695006.1 | |
XM_017001020.1 | 1254 | Intron | XP_016856509.1 | |||
XM_017001021.1 | 1254 | Intron | XP_016856510.1 | |||
XM_017001022.1 | 1254 | Missense Mutation | CGG,CTG | R,L 402 | XP_016856511.1 | |
XM_017001023.1 | 1254 | Missense Mutation | CGG,CTG | R,L 402 | XP_016856512.1 |
RPS8 - ribosomal protein S8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD38A - small nucleolar RNA, C/D box 38A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD38B - small nucleolar RNA, C/D box 38B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD46 - small nucleolar RNA, C/D box 46 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD55 - small nucleolar RNA, C/D box 55 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |