Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCATTCTTGAAAACCTATCTCA[A/G]GAATCACTTCCTTTAGTATTAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606357 MIM: 609367 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX21 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDX21 - DEAD-box helicase 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIF1BP - KIF1 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015634.3 | Intron | NP_056449.1 | ||||
XM_017016067.1 | Intron | XP_016871556.1 |