Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAAGGGGCTGCGCGGCTGCTTGG[A/G]CCACTTGTTATTTTTTGCGCACAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610679 | ||||||||||||||||||||
Literature Links: |
CDK14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK14 - cyclin dependent kinase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287135.1 | Intron | NP_001274064.1 | ||||
NM_001287136.1 | Intron | NP_001274065.1 | ||||
NM_001287137.1 | Intron | NP_001274066.1 | ||||
NM_012395.3 | Intron | NP_036527.1 | ||||
XM_005250436.3 | Intron | XP_005250493.1 | ||||
XM_005250438.3 | Intron | XP_005250495.1 | ||||
XM_005250439.2 | Intron | XP_005250496.1 | ||||
XM_011516306.2 | Intron | XP_011514608.1 | ||||
XM_011516307.2 | Intron | XP_011514609.1 | ||||
XM_017012320.1 | Intron | XP_016867809.1 | ||||
XM_017012321.1 | Intron | XP_016867810.1 | ||||
XM_017012322.1 | Intron | XP_016867811.1 |