Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATAAATGCCCTATTTATGTGTGA[A/C]ACAAATTAGGTTCTATTTTTCAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605094 | ||||||||||||||||||||
Literature Links: |
STEAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STEAP2 - STEAP2 metalloreductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040665.1 | 2857 | Intron | NP_001035755.1 | |||
NM_001040666.1 | 2857 | Intron | NP_001035756.1 | |||
NM_001244944.1 | 2857 | Intron | NP_001231873.1 | |||
NM_001244945.1 | 2857 | Intron | NP_001231874.1 | |||
NM_001244946.1 | 2857 | Intron | NP_001231875.1 | |||
NM_152999.3 | 2857 | Intron | NP_694544.2 | |||
XM_006715921.3 | 2857 | Intron | XP_006715984.1 | |||
XM_017011953.1 | 2857 | Intron | XP_016867442.1 | |||
XM_017011954.1 | 2857 | Intron | XP_016867443.1 | |||
XM_017011955.1 | 2857 | Intron | XP_016867444.1 | |||
XM_017011956.1 | 2857 | Intron | XP_016867445.1 | |||
XM_017011957.1 | 2857 | Intron | XP_016867446.1 | |||
XM_017011958.1 | 2857 | UTR 5 | XP_016867447.1 |
STEAP2-AS1 - STEAP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |