Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATGGGGCCTCCGTGAGTGAGAT[C/T]CGTCATAGTGCGCTGCAGCGATGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613653 MIM: 616302 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP5Z1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AP5Z1 - adaptor related protein complex 5 zeta 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014855.2 | Intron | NP_055670.1 | ||||
XM_017012864.1 | Intron | XP_016868353.1 |
FOXK1 - forkhead box K1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4656 - microRNA 4656 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |