Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCCCACCCCACCTCCCCACAGC[C/T]TTCTGTAAAGGTCAGAGCAAATTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604630 MIM: 610325 | ||||||||||||||||||||
Literature Links: |
NR0B2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NR0B2 - nuclear receptor subfamily 0 group B member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021969.2 | Intron | NP_068804.1 | ||||
XM_011542297.2 | Intron | XP_011540599.1 |
NUDC - nuclear distribution C, dynein complex regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006600.3 | Intron | NP_006591.1 | ||||
XM_011540529.1 | Intron | XP_011538831.1 | ||||
XM_011540530.1 | Intron | XP_011538832.1 | ||||
XM_017000094.1 | Intron | XP_016855583.1 | ||||
XM_017000095.1 | Intron | XP_016855584.1 | ||||
XM_017000096.1 | Intron | XP_016855585.1 |