Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATAATAAAATTGTACACTTTGG[C/T]ATGTGAATTATATCTCCATGAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616070 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC113 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC113 - coiled-coil domain containing 113 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142302.1 | Intron | NP_001135774.1 | ||||
NM_014157.3 | Intron | NP_054876.2 | ||||
XM_011523038.2 | Intron | XP_011521340.1 | ||||
XM_011523039.2 | Intron | XP_011521341.1 | ||||
XM_011523040.2 | Intron | XP_011521342.1 | ||||
XM_011523041.2 | Intron | XP_011521343.1 | ||||
XM_011523042.2 | Intron | XP_011521344.1 | ||||
XM_017023176.1 | Intron | XP_016878665.1 | ||||
XM_017023177.1 | Intron | XP_016878666.1 | ||||
XM_017023178.1 | Intron | XP_016878667.1 |