Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTACGTTTCCACAAGCTGACTT[G/T]GGTTGGGAGCTCATCTGTTCCCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608943 | ||||||||||||||||||||
Literature Links: |
CIAPIN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CIAPIN1 - cytokine induced apoptosis inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308347.1 | Intron | NP_001295276.1 | ||||
NM_001308358.1 | Intron | NP_001295287.1 | ||||
NM_020313.3 | Intron | NP_064709.2 | ||||
XM_005256063.4 | Intron | XP_005256120.1 |