Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAAATTTTTCAGAAGAGCTTAC[A/G]ATTAGATGAACTAAAAGAACTAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613222 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNPDA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GNPDA2 - glucosamine-6-phosphate deaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270880.1 | Intron | NP_001257809.1 | ||||
NM_001270881.1 | Intron | NP_001257810.1 | ||||
NM_138335.2 | Intron | NP_612208.1 |
GUF1 - GUF1 homolog, GTPase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |