Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGGAAAACTCTACAGTATAGAC[A/T]TCAATAGAAAACAATTTCCAATCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614248 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEAK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PEAK1 - pseudopodium enriched atypical kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024776.3 | 8492 | UTR 3 | NP_079052.2 | |||
XM_005254670.2 | 8492 | UTR 3 | XP_005254727.1 | |||
XM_005254671.2 | 8492 | UTR 3 | XP_005254728.1 | |||
XM_005254673.2 | 8492 | UTR 3 | XP_005254730.1 | |||
XM_005254674.2 | 8492 | UTR 3 | XP_005254731.1 | |||
XM_005254675.4 | 8492 | UTR 3 | XP_005254732.1 | |||
XM_011522033.2 | 8492 | UTR 3 | XP_011520335.1 | |||
XM_011522034.2 | 8492 | UTR 3 | XP_011520336.1 | |||
XM_011522035.2 | 8492 | UTR 3 | XP_011520337.1 | |||
XM_011522036.2 | 8492 | UTR 3 | XP_011520338.1 | |||
XM_011522037.2 | 8492 | UTR 3 | XP_011520339.1 | |||
XM_011522038.2 | 8492 | UTR 3 | XP_011520340.1 | |||
XM_011522039.2 | 8492 | UTR 3 | XP_011520341.1 | |||
XM_011522040.1 | 8492 | Intron | XP_011520342.1 | |||
XM_011522041.1 | 8492 | Intron | XP_011520343.1 | |||
XM_011522042.2 | 8492 | UTR 3 | XP_011520344.1 |
Set Membership: |
HapMap |