Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGTTTCCGGAAGCAGCTTGAGT[C/T]CGGACACCAACGACGGGGACGCTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610347 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C18orf21 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C18orf21 - chromosome 18 open reading frame 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201474.1 | 10 | UTR 5 | NP_001188403.1 | |||
NM_001201475.1 | 10 | UTR 5 | NP_001188404.1 | |||
NM_001201476.1 | 10 | UTR 5 | NP_001188405.1 | |||
NM_031446.4 | 10 | UTR 5 | NP_113634.3 | |||
XM_005258364.4 | 10 | Intron | XP_005258421.1 |
RPRD1A - regulation of nuclear pre-mRNA domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |