Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGAACTAATAAAAATATACCTCC[A/G]TTAAACTGCTCAGACTCAAGAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF519 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF519 - zinc finger protein 519 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145287.3 | 16239 | Intron | NP_660330.2 | |||
XM_011525605.2 | 16239 | Intron | XP_011523907.1 | |||
XM_017025562.1 | 16239 | UTR 3 | XP_016881051.1 | |||
XM_017025563.1 | 16239 | UTR 3 | XP_016881052.1 | |||
XM_017025564.1 | 16239 | UTR 3 | XP_016881053.1 | |||
XM_017025565.1 | 16239 | UTR 3 | XP_016881054.1 | |||
XM_017025566.1 | 16239 | Intron | XP_016881055.1 |