Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGGCCAGATCGCTCTTCCAGCGC[A/G]GGTTGGTCCACCAGGCAGAGCCCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165190 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FGF5 - fibroblast growth factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291812.1 | 31 | UTR 5 | NP_001278741.1 | |||
NM_004464.3 | 31 | Intron | NP_004455.2 | |||
NM_033143.2 | 31 | Intron | NP_149134.1 |
Set Membership: |
HapMap |