Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTAAATAGGTAAGGCCACTTTAA[A/T]CTTTATCAGGTATATCTATCGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159405 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYBL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MYBL1 - MYB proto-oncogene like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080416.3 | Intron | NP_001073885.1 | ||||
NM_001144755.2 | Intron | NP_001138227.1 | ||||
NM_001294282.1 | Intron | NP_001281211.1 | ||||
XM_011517533.2 | Intron | XP_011515835.1 | ||||
XM_011517535.2 | Intron | XP_011515837.1 | ||||
XM_017013455.1 | Intron | XP_016868944.1 | ||||
XM_017013456.1 | Intron | XP_016868945.1 | ||||
XM_017013457.1 | Intron | XP_016868946.1 | ||||
XM_017013458.1 | Intron | XP_016868947.1 | ||||
XM_017013459.1 | Intron | XP_016868948.1 |
Set Membership: |
HapMap |