Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCACTGAGGCCTGTGTATGCAAC[A/G]TATCAGTGAACAGTTGCATGACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610972 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AJAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AJAP1 - adherens junctions associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042478.1 | Intron | NP_001035943.1 | ||||
NM_018836.3 | Intron | NP_061324.1 | ||||
XM_011541786.2 | Intron | XP_011540088.1 | ||||
XM_011541787.2 | Intron | XP_011540089.1 |
Set Membership: |
HapMap |